missing translation for 'onlineSavingsMsg'
Learn More

Sulfaguanidine 98.0+%, TCI America™

Catalog No. S046425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S046425G 25 g
S0464500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S046425G Supplier TCI America Supplier No. S046425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57-67-0
Molecular Formula C7H10N4O2S
Molecular Weight (g/mol) 214.243
MDL Number MFCD00038136
InChI Key BRBKOPJOKNSWSG-UHFFFAOYSA-N
Synonym sulfaguanidine, sulphaguanidine, sulfaguanidin, guanicil, sulfaguine, aterian, sulfanilguanidine, sulfanilylguanidine, sulfoguanidine, abiguanil
PubChem CID 5324
IUPAC Name 2-(4-aminophenyl)sulfonylguanidine
SMILES C1=CC(=CC=C1N)S(=O)(=O)N=C(N)N

Specifications

Melting Point 189°C
Color White
Quantity 25 g
Formula Weight 214.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sulfaguanidine
EINECSNumber (3)-1977
RTECSNumber WO8575000
TSCA Yes