missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfadiazine Sodium USP, Medisca™
$138.00 - $12163.07
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
18-048-055
|
Medisca Pharmaceutique Inc
322807** |
25 kg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18-048-096
|
Medisca Pharmaceutique Inc
322808* |
500 g |
Each for $401.76
|
|
|||||
|
18-048-097
|
Medisca Pharmaceutique Inc
322803 |
5 kg |
Each for $2,686.54
|
|
|||||
|
18-048-098
|
Medisca Pharmaceutique Inc
322805* |
100 g |
Each for $138.00
|
|
|||||
|
18-048-115
|
Medisca Pharmaceutique Inc
322801 |
2.5 kg |
Each for $1,416.87
|
|
|||||
|
18-048-140
|
Medisca Pharmaceutique Inc
322809 |
1 kg |
Each for $720.72
|
|
|||||
Description
Sulfadiazine Sodium USP Not for Human Use.
Chemical Identifiers
| 547-32-0 |
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 547-32-0 | |
| 25 kg | |
| USP |
| Crystalline Powder | |
| White | |
| 99.0 to 100.5% |