missing translation for 'onlineSavingsMsg'
Learn More

Sulfadiazine sodium salt, ≥98% (HPLC), MilliporeSigma™ Supelco™

$56.61 - $171.36

Specifications

CAS 547-32-0
Physical Form Crystalline Powder
Linear Formula C10H9N4 NaO2 S
MDL Number MFCD00067333
Synonym 4-Amino-N-(2-pyrimidinyl)benzenesulfonamide sodium salt; SPS Agar Supplement
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111014573
View Documents
Merck Emd Millipore
S638725G
25 g
Each for $56.61
Only null left
 
111014574
View Documents
Merck Emd Millipore
S6387100G
100 g
Each for $171.36
Only null left
 
Description

Description

Sulfadiazine sodium salt is a broad-spectrum antibiotic belonging to the class of sulfonamides commonly used against gram-postive and gram-negative microorganisms in humans and veterinary medication. Its mode of action involves inhibiting the multiplication of bacteria as competitive inhibitors of p-aminobenzoic acid in the folic acid metabolism cycle. Featured Industries: Pharmaceutical (small molecule)
Specifications

Specifications

547-32-0
C10H9N4 NaO2 S
4-Amino-N-(2-pyrimidinyl)benzenesulfonamide sodium salt; SPS Agar Supplement
272.26
Crystalline Powder
MFCD00067333
Organic Standard
≥98% (HPLC)
Videos
SDS
Documents

Documents