missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfadiazine sodium salt, ≥98% (HPLC), MilliporeSigma™ Supelco™
Description
Sulfadiazine sodium salt is a broad-spectrum antibiotic belonging to the class of sulfonamides commonly used against gram-postive and gram-negative microorganisms in humans and veterinary medication. Its mode of action involves inhibiting the multiplication of bacteria as competitive inhibitors of p-aminobenzoic acid in the folic acid metabolism cycle. Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 547-32-0 |
| Physical Form | Crystalline Powder |
| Linear Formula | C10H9N4 NaO2 S |
| MDL Number | MFCD00067333 |
| Synonym | 4-Amino-N-(2-pyrimidinyl)benzenesulfonamide sodium salt; SPS Agar Supplement |
| Type | Organic Standard |
| Quantity | 100 g |
| Formula Weight | 272.26 |
| Percent Purity | ≥98% (HPLC) |