missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfadiazine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 68-35-9 |
| Physical Form | Neat |
| Linear Formula | C10H10N4O2 S |
| MDL Number | MFCD00006065 |
| Melting Point | 253°C |
| Synonym | Sulfadiazine; 4-Amino-N-(2-pyrimidinyl)benzenesulfonamide; N1-(Pyrimidin-2-yl)sulfanilamide |
| Type | Certified Reference Material |
| Quantity | 500 mg |
| Formula Weight | 250.28 |
| Grade | Certified Reference Material |