missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfacetamide 98.0+%, TCI America™
Supplier: TCI America S057725G
Specifications
| Sulfacetamide | |
| 183°C | |
| C8H10N2O3S | |
| sulfacetamide, sulphacetamide, acetosulfamine, n-acetylsulfanilamide, acetosulfamin, albucid, n-sulfanilylacetamide, n-4-aminophenyl sulfonyl acetamide, sulfacetimide, acetocid | |
| CC(=O)NS(=O)(=O)C1=CC=C(C=C1)N | |
| 214.239 | |
| CHEBI:63845 | |
| ≥98.0% (HPLC,T) |
| 144-80-9 | |
| White | |
| 25 g | |
| SKIVFJLNDNKQPD-UHFFFAOYSA-N | |
| N-(4-aminophenyl)sulfonylacetamide | |
| 5320 | |
| 214.24 | |
| Crystalline Powder |
Chemical Identifiers
| 144-80-9 | |
| 214.239 | |
| sulfacetamide, sulphacetamide, acetosulfamine, n-acetylsulfanilamide, acetosulfamin, albucid, n-sulfanilylacetamide, n-4-aminophenyl sulfonyl acetamide, sulfacetimide, acetocid | |
| CHEBI:63845 | |
| CC(=O)NS(=O)(=O)C1=CC=C(C=C1)N |
| C8H10N2O3S | |
| SKIVFJLNDNKQPD-UHFFFAOYSA-N | |
| 5320 | |
| N-(4-aminophenyl)sulfonylacetamide |
Safety and Handling
RTECSNumber : AC8450000
TSCA : Yes