missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfacetamide 98.0+%, TCI America™
$44.73 - $301.98
Chemical Identifiers
| CAS | 144-80-9 |
|---|---|
| Molecular Formula | C8H10N2O3S |
| Molecular Weight (g/mol) | 214.239 |
| InChI Key | SKIVFJLNDNKQPD-UHFFFAOYSA-N |
| Synonym | sulfacetamide, sulphacetamide, acetosulfamine, n-acetylsulfanilamide, acetosulfamin, albucid, n-sulfanilylacetamide, n-4-aminophenyl sulfonyl acetamide, sulfacetimide, acetocid |
| PubChem CID | 5320 |
| ChEBI | CHEBI:63845 |
| IUPAC Name | N-(4-aminophenyl)sulfonylacetamide |
| SMILES | CC(=O)NS(=O)(=O)C1=CC=C(C=C1)N |
Chemical Identifiers
| 144-80-9 | |
| 214.239 | |
| sulfacetamide, sulphacetamide, acetosulfamine, n-acetylsulfanilamide, acetosulfamin, albucid, n-sulfanilylacetamide, n-4-aminophenyl sulfonyl acetamide, sulfacetimide, acetocid | |
| CHEBI:63845 | |
| CC(=O)NS(=O)(=O)C1=CC=C(C=C1)N |
| C8H10N2O3S | |
| SKIVFJLNDNKQPD-UHFFFAOYSA-N | |
| 5320 | |
| N-(4-aminophenyl)sulfonylacetamide |
Specifications
| 144-80-9 | |
| White | |
| 25 g | |
| SKIVFJLNDNKQPD-UHFFFAOYSA-N | |
| N-(4-aminophenyl)sulfonylacetamide | |
| 5320 | |
| 214.24 | |
| Crystalline Powder |
| 183°C | |
| C8H10N2O3S | |
| sulfacetamide, sulphacetamide, acetosulfamine, n-acetylsulfanilamide, acetosulfamin, albucid, n-sulfanilylacetamide, n-4-aminophenyl sulfonyl acetamide, sulfacetimide, acetocid | |
| CC(=O)NS(=O)(=O)C1=CC=C(C=C1)N | |
| 214.239 | |
| CHEBI:63845 | |
| ≥98.0% (HPLC,T) | |
| Sulfacetamide |
Safety and Handling
RTECSNumber : AC8450000
TSCA : Yes