missing translation for 'onlineSavingsMsg'
Learn More

Sulfacetamide Sodium Salt Hydrate 98.0+%, TCI America™

Catalog No. p-7135927
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S057825G 25 g
S0578250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S057825G Supplier TCI America Supplier No. S057825G
Only null left

Chemical Identifiers

CAS 127-56-0
Molecular Formula C8H9N2NaO3S
Molecular Weight (g/mol) 236.22
MDL Number MFCD00149555
InChI Key PQMSFAORUFMASU-UHFFFAOYSA-M
Synonym N-Acetyl-4-aminobenzenesulfonamide Sodium Salt, N-Acetylsulfanilamide Sodium Salt
PubChem CID 4022878
ChEBI CHEBI:9327
IUPAC Name sodium acetyl(4-aminobenzenesulfonyl)azanide
SMILES [Na+].CC(=O)[N-]S(=O)(=O)C1=CC=C(N)C=C1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 236.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sulfacetamide Sodium Salt Hydrate
EINECSNumber (9)-0038
RTECSNumber AC8480000
TSCA Yes