missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfacetamide Sodium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 6209-17-2 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C8H9N2O3 SNa |
| MDL Number | MFCD00007885 |
| Synonym | Sulfacetamide sodium salt; N-(p-Aminobenzenesulfonyl)acetamide |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 236.22 |
| Grade | Certified Reference Material |