missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfacetamide Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 144-80-9 |
| Linear Formula | C8H10N2O3 S |
| MDL Number | MFCD00066501 |
| Synonym | Sulfacetamide; N-(4-Aminobenzenesulfonyl)acetamide; N-(4-Aminophenylsulfonyl)acetamide; N1-Acetylsulfanilamide |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 214.24 |
| Grade | Certified Reference Material |