missing translation for 'onlineSavingsMsg'
Learn More

Sulbactam 98.0+%, TCI America™

Catalog No. p-7135973
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08685G 5 g
S086825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08685G Supplier TCI America Supplier No. S08685G
Only null left

Chemical Identifiers

CAS 68373-14-8
Molecular Formula C8H11NO5S
Molecular Weight (g/mol) 233.24
MDL Number MFCD00867005
InChI Key FKENQMMABCRJMK-RITPCOANSA-N
Synonym sulbactam, sulbactam acid, penicillanic acid sulfone, sulbactamum, betamaze, 2s,5r-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid 4,4-dioxide, penicillanic acid 1,1-dioxide, unii-s4tf6i2330, penicillanic acid dioxide, chembl403
PubChem CID 130313
ChEBI CHEBI:9321
IUPAC Name (2S,5R)-3,3-dimethyl-4,4,7-trioxo-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMILES CC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O

Specifications

Melting Point 156°C
Color White-Yellow
Quantity 5 g
Formula Weight 233.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sulbactam
TSCA No
Recommended Storage Freezer