Learn More
Sulbactam, 98%
An antibacterial agent used in the treatment of multidrug resistant bacterium, Acinetobacter, a nosocomial pathogen. Also used in cell signaling studies. | CAS: 68373-14-8 | C8H11NO5S | 233.24 g/mol
Supplier: Thermo Scientific Chemicals J6623506
Description
Sulbactam is an antibacterial agent used in the treatment of multidrug resistant bacterium, Acinetobacter, a nosocomial pathogen. Also used in cell signaling studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Sulbactam | |
| 154°C to 157°C | |
| C8H11NO5S | |
| 5 g | |
| Soluble in water at 18mg/ml | |
| CC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O | |
| 233.24 | |
| CHEBI:9321 | |
| 98% |
| 68373-14-8 | |
| Glass bottle | |
| MFCD00867005 | |
| sulbactam, sulbactam acid, penicillanic acid sulfone, sulbactamum, betamaze, 2s,5r-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid 4,4-dioxide, penicillanic acid 1,1-dioxide, unii-s4tf6i2330, penicillanic acid dioxide, chembl403 | |
| FKENQMMABCRJMK-RITPCOANSA-N | |
| (2S,5R)-3,3-dimethyl-4,4,7-trioxo-4λâ¶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid | |
| 130313 | |
| 233.24 | |
| Powder |
Chemical Identifiers
| 68373-14-8 | |
| 233.24 | |
| FKENQMMABCRJMK-RITPCOANSA-N | |
| 130313 | |
| (2S,5R)-3,3-dimethyl-4,4,7-trioxo-4λâ¶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid |
| C8H11NO5S | |
| MFCD00867005 | |
| sulbactam, sulbactam acid, penicillanic acid sulfone, sulbactamum, betamaze, 2s,5r-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid 4,4-dioxide, penicillanic acid 1,1-dioxide, unii-s4tf6i2330, penicillanic acid dioxide, chembl403 | |
| CHEBI:9321 | |
| CC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 269-878-2
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only