missing translation for 'onlineSavingsMsg'
Learn More

Sulbactam, 98%

Catalog No. AAJ6623503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6623503 1 g
AAJ6623506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6623503 Supplier Thermo Scientific Chemicals Supplier No. J6623503

An antibacterial agent used in the treatment of multidrug resistant bacterium, Acinetobacter, a nosocomial pathogen. Also used in cell signaling studies. | CAS: 68373-14-8 | C8H11NO5S | 233.24 g/mol

Sulbactam is an antibacterial agent used in the treatment of multidrug resistant bacterium, Acinetobacter, a nosocomial pathogen. Also used in cell signaling studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68373-14-8
Molecular Formula C8H11NO5S
Molecular Weight (g/mol) 233.24
MDL Number MFCD00867005
InChI Key FKENQMMABCRJMK-RITPCOANSA-N
Synonym sulbactam, sulbactam acid, penicillanic acid sulfone, sulbactamum, betamaze, 2s,5r-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid 4,4-dioxide, penicillanic acid 1,1-dioxide, unii-s4tf6i2330, penicillanic acid dioxide, chembl403
PubChem CID 130313
ChEBI CHEBI:9321
IUPAC Name (2S,5R)-3,3-dimethyl-4,4,7-trioxo-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMILES CC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O

Specifications

Melting Point 154°C to 157°C
Packaging Glass bottle
Quantity 1 g
Solubility Information Soluble in water at 18mg/ml
Formula Weight 233.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sulbactam
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 269-878-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only