missing translation for 'onlineSavingsMsg'
Learn More

Sudan orange G, pure

CAS: 2051-85-6 | C12H10N2O2 | 214.22 g/mol

$54.29 - $171.80

Chemical Identifiers

CAS 2051-85-6
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002275
InChI Key PULNESMFDLBKAZ-SDNWHVSQSA-N
Synonym C.I. 11920, 4-(Phenylazo)resorcinol, Solvent orange 1
PubChem CID 5921655
IUPAC Name (4Z)-3-hydroxy-4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one
SMILES OC1=CC(=O)C=C\C1=N/NC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC190170050
SDS View Documents
Thermo Scientific Chemicals
190170050
5 g Glass bottle
Each for $54.29
Only null left
 
AC190170250
SDS View Documents
Thermo Scientific Chemicals
190170250
25 g Glass bottle
Each for $171.80
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2051-85-6
214.22
PULNESMFDLBKAZ-SDNWHVSQSA-N
5921655
OC1=CC(=O)C=C\C1=N/NC1=CC=CC=C1
C12H10N2O2
MFCD00002275
C.I. 11920, 4-(Phenylazo)resorcinol, Solvent orange 1
(4Z)-3-hydroxy-4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one

Specifications

2051-85-6
MFCD00002275
PULNESMFDLBKAZ-SDNWHVSQSA-N
OC1=CC(=O)C=C\C1=N/NC1=CC=CC=C1
214.22
214.22
Glass bottle
Orange to Red
Powder
C12H10N2O2
C.I. 11920, 4-(Phenylazo)resorcinol, Solvent orange 1
Authentic
(4Z)-3-hydroxy-4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one
5921655
Extra Pure
Complies
5 g
Sudan orange G, Pure
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 218-131-9

RTECSNumber : CZ9027500

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only