missing translation for 'onlineSavingsMsg'
Learn More

Sudan orange G, pure

CAS: 2051-85-6 | C12H10N2O2 | 214.22 g/mol

Supplier:  Thermo Scientific Chemicals 190170050

 View more versions of this product

Catalog No. AC190170050


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sudan orange G
2051-85-6
MFCD00002275
PULNESMFDLBKAZ-SDNWHVSQSA-N
OC1=CC(=O)C=C\C1=N/NC1=CC=CC=C1
214.22
214.22
Glass bottle
Orange to Red
Powder
Pure
C12H10N2O2
C.I. 11920, 4-(Phenylazo)resorcinol, Solvent orange 1
Authentic
(4Z)-3-hydroxy-4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one
5921655
Extra Pure
Complies
5 g

Chemical Identifiers

2051-85-6
214.22
PULNESMFDLBKAZ-SDNWHVSQSA-N
5921655
OC1=CC(=O)C=C\C1=N/NC1=CC=CC=C1
C12H10N2O2
MFCD00002275
C.I. 11920, 4-(Phenylazo)resorcinol, Solvent orange 1
(4Z)-3-hydroxy-4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 218-131-9

RTECSNumber : CZ9027500

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only