missing translation for 'onlineSavingsMsg'
Learn More

Sudan Orange G, ≥96.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013485
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013485 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013485 Supplier Merck Emd Millipore Supplier No. 4320725MG
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 2051-85-6
Physical Form Neat
Linear Formula C6H5 N=NC6H3-1,3-(OH)2
MDL Number MFCD00002275
Melting Point 143°C to 146°C
Synonym 2,4-Dihydroxyazobenzene; 4-(Phenylazo)resorcinol; SOG
Type Analytical Standard
Quantity 25 mg
Formula Weight 214.22
Percent Purity ≥96.0% (HPLC)
Grade Analytical Standard
Show More Show Less