missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sudan IV-d6, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 1014689-18-9 |
| Physical Form | Neat |
| Linear Formula | C24D6H14N4 O |
| MDL Number | MFCD08460518 |
| Synonym | 1-[2-Methyl-4-(2-methyl-phenylazo)phenylazo]-2-naphthol-d6 |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 386.48 |
| Grade | Analytical Standard |