Learn More
Thermo Scientific Chemicals Sudan IV
CAS: 85-83-6 | C24H20N4O | 380.45 g/mol
$68.85 - $165.05
Chemical Identifiers
| CAS | 85-83-6 |
|---|---|
| Molecular Formula | C24H20N4O |
| Molecular Weight (g/mol) | 380.45 |
| MDL Number | MFCD00003893 |
| InChI Key | KMDLOETUWUPGMB-BXCCFQQFSA-N |
| Synonym | C.I. 26105; Scarlet Red |
| PubChem CID | 6797604 |
| IUPAC Name | (1Z)-1-(2-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-1,2-dihydronaphthalen-2-one |
| SMILES | CC1=CC(=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12)N=NC1=CC=CC=C1C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1218114
|
Thermo Scientific Chemicals
A1218114 |
25 g |
Each for $68.85
|
|
|||||
|
AAA1218122
|
Thermo Scientific Chemicals
A1218122 |
100 g |
Each for $165.05
|
|
|||||
Description
Useful as a stain for fat in animal tissue
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 85-83-6 | |
| 380.45 | |
| KMDLOETUWUPGMB-BXCCFQQFSA-N | |
| 6797604 | |
| CC1=CC(=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12)N=NC1=CC=CC=C1C |
| C24H20N4O | |
| MFCD00003893 | |
| C.I. 26105; Scarlet Red | |
| (1Z)-1-(2-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-1,2-dihydronaphthalen-2-one |
Specifications
| ∼185°C (decomposition) | |
| C24H20N4O | |
| 14,8393 | |
| KMDLOETUWUPGMB-BXCCFQQFSA-N | |
| (1Z)-1-(2-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-1,2-dihydronaphthalen-2-one | |
| 6797604 | |
| 25 g |
| 85-83-6 | |
| MFCD00003893 | |
| C.I. 26105; Scarlet Red | |
| CC1=CC(=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12)N=NC1=CC=CC=C1C | |
| 380.45 | |
| 380.45 | |
| Sudan IV |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 201-635-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only