missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan IV

CAS: 85-83-6 | C24H20N4O | 380.45 g/mol

Supplier:  Thermo Scientific Chemicals A1218122

 View more versions of this product

Catalog No. AAA1218122


Only null left
Explore available promotions

Description

Description

Useful as a stain for fat in animal tissue

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sudan IV
85-83-6
MFCD00003893
C.I. 26105; Scarlet Red
CC1=CC(=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12)N=NC1=CC=CC=C1C
380.45
380.45
∼185°C (decomposition)
C24H20N4O
14,8393
KMDLOETUWUPGMB-BXCCFQQFSA-N
(1Z)-1-(2-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-1,2-dihydronaphthalen-2-one
6797604
100 g

Chemical Identifiers

85-83-6
380.45
KMDLOETUWUPGMB-BXCCFQQFSA-N
6797604
CC1=CC(=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12)N=NC1=CC=CC=C1C
C24H20N4O
MFCD00003893
C.I. 26105; Scarlet Red
(1Z)-1-(2-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-1,2-dihydronaphthalen-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 201-635-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only