missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sudan III 90.0+%, TCI America™
$48.46 - $143.54
Chemical Identifiers
| CAS | 85-86-9 |
|---|---|
| Molecular Formula | C22H16N4O |
| Molecular Weight (g/mol) | 352.397 |
| MDL Number | MFCD00003905 |
| InChI Key | HTPQPMPFXUWUOT-UHFFFAOYSA-N |
| Synonym | sudan iii, c.i. solvent red 23, 2-naphthalenol, 1-4-phenylazo phenyl azo, fhninjwbtrxebc-qszpnpogsa-n, sudaniii, sudan iiisolvent red 23, 2-naphthalenol, 1-2-4-2-phenyldiazenyl phenyl diazenyl, 1-2-4-2-phenyldiazen-1-yl phenyl diazen-1-yl naphthalen-2-ol, 1-e-4-e-phenylazo phenyl azo naphthalen-2-ol, 1-4-phenyldiazenylphenyl hydrazinylidene naphthalen-2-one |
| PubChem CID | 6789251 |
| IUPAC Name | 1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one |
| SMILES | C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43 |
Chemical Identifiers
| 85-86-9 | |
| 352.397 | |
| HTPQPMPFXUWUOT-UHFFFAOYSA-N | |
| 6789251 | |
| C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43 |
| C22H16N4O | |
| MFCD00003905 | |
| sudan iii, c.i. solvent red 23, 2-naphthalenol, 1-4-phenylazo phenyl azo, fhninjwbtrxebc-qszpnpogsa-n, sudaniii, sudan iiisolvent red 23, 2-naphthalenol, 1-2-4-2-phenyldiazenyl phenyl diazenyl, 1-2-4-2-phenyldiazen-1-yl phenyl diazen-1-yl naphthalen-2-ol, 1-e-4-e-phenylazo phenyl azo naphthalen-2-ol, 1-4-phenyldiazenylphenyl hydrazinylidene naphthalen-2-one | |
| 1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one |
Specifications
| 85-86-9 | |
| C22H16N4O | |
| 25 g | |
| HTPQPMPFXUWUOT-UHFFFAOYSA-N | |
| 1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one | |
| 6789251 | |
| ≥90.0% (E) | |
| Sudan III |
| 202°C | |
| MFCD00003905 | |
| sudan iii, c.i. solvent red 23, 2-naphthalenol, 1-4-phenylazo phenyl azo, fhninjwbtrxebc-qszpnpogsa-n, sudaniii, sudan iiisolvent red 23, 2-naphthalenol, 1-2-4-2-phenyldiazenyl phenyl diazenyl, 1-2-4-2-phenyldiazen-1-yl phenyl diazen-1-yl naphthalen-2-ol, 1-e-4-e-phenylazo phenyl azo naphthalen-2-ol, 1-4-phenyldiazenylphenyl hydrazinylidene naphthalen-2-one | |
| C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43 | |
| 352.397 | |
| 352.40 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-3087
RTECSNumber : QK4250000
TSCA : Yes