Learn More
Thermo Scientific Chemicals Sudan I, pure
CAS: 842-07-9 | C16H12N2O | 248.285 g/mol
$76.28 - $133.78
Chemical Identifiers
| CAS | 842-07-9 |
|---|---|
| Molecular Formula | C16H12N2O |
| Molecular Weight (g/mol) | 248.285 |
| MDL Number | MFCD00003911 |
| InChI Key | ZONYAPYTDIVJGG-VLGSPTGOSA-N |
| Synonym | C.I. 12055, 1-Phenylazo-2-naphthol, Solvent Yellow 14 |
| PubChem CID | 5858445 |
| IUPAC Name | (1Z)-1-(phenylhydrazinylidene)naphthalen-2-one |
| SMILES | C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC=CC=C32 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC147060250
|
Thermo Scientific Chemicals
147060250 |
25 g | Glass bottle |
Each for $76.28
|
|
||||
|
AC147061000
|
Thermo Scientific Chemicals
147061000 |
100 g | Glass bottle |
Each for $133.78
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 842-07-9 | |
| 248.285 | |
| ZONYAPYTDIVJGG-VLGSPTGOSA-N | |
| 5858445 | |
| C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC=CC=C32 |
| C16H12N2O | |
| MFCD00003911 | |
| C.I. 12055, 1-Phenylazo-2-naphthol, Solvent Yellow 14 | |
| (1Z)-1-(phenylhydrazinylidene)naphthalen-2-one |
Specifications
| 131.0°C to 133.0°C | |
| C16H12N2O | |
| 16, IV, 228 | |
| ZONYAPYTDIVJGG-VLGSPTGOSA-N | |
| C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC=CC=C32 | |
| 248.285 | |
| at approximately 418 and 476nm (in ethanol) | |
| Pure | |
| 500 min. (at 476nm, in ethanol) | |
| 25 g | |
| Sudan I |
| 842-07-9 | |
| MFCD00003911 | |
| C.I. 12055, 1-Phenylazo-2-naphthol, Solvent Yellow 14 | |
| Authentic | |
| (1Z)-1-(phenylhydrazinylidene)naphthalen-2-one | |
| 5858445 | |
| 248.28 | |
| Glass bottle | |
| Brown or Orange to Red | |
| Powder |
Safety and Handling
GHS H Statement:
Suspected of causing genetic defects.
Suspected of causing cancer.
May cause an allergic skin reaction.
May cause long lasting harmful effects to aquatic life.
GHS P Statement:
Wear eye protection/face protection.
Avoid release to the environment.
Avoid breathing dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CANCER SUSPECTED AGENT
EINECSNumber : 212-668-2
RTECSNumber : QL4900000
TSCA : TSCA
RUO – Research Use Only