missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sudan I, pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 131.0°C to 133.0°C |
| Beilstein | 16, IV, 228 |
| Infrared Spectrum | Authentic |
| Lambda Max | at approximately 418 and 476nm (in ethanol) |
| Formula Weight | 248.28 |
| Grade | Pure |
| Packaging | Glass bottle |
| Specific Extinction | 500 min. (at 476nm, in ethanol) |
| Color | Brown or Orange to Red |
| Quantity | 100 g |
| Show More |
Safety and Handling
| Hazard Category | CANCER SUSPECTED AGENT |
| Hazard Statement | GHS H Statement: Suspected of causing genetic defects. Suspected of causing cancer. May cause an allergic skin reaction. May cause long lasting harmful effects to aquatic life. |
| Precautionary Statement | GHS P Statement: Wear eye protection/face protection. Avoid release to the environment. Avoid breathing dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 212-668-2 |
| RTECSNumber | QL4900000 |
| TSCA | TSCA |
RUO – Research Use Only