missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sudan Black B, high purity biological stain
CAS: 4197-25-5 | C29H24N6 | 456.553 g/mol
$161.45
Chemical Identifiers
| CAS | 4197-25-5 |
|---|---|
| Molecular Formula | C29H24N6 |
| Molecular Weight (g/mol) | 456.553 |
| MDL Number | MFCD00006919 |
| InChI Key | YCUVUDODLRLVIC-UHFFFAOYSA-N |
| Synonym | C.I. 26150, Fat Black HB, Solvent Black 3 |
| PubChem CID | 61336 |
| IUPAC Name | (2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene |
| SMILES | CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC190160250
|
Thermo Scientific Chemicals
190160250 |
25 g | Glass bottle |
Each for $161.45
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4197-25-5 | |
| 456.553 | |
| YCUVUDODLRLVIC-UHFFFAOYSA-N | |
| 61336 | |
| CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C |
| C29H24N6 | |
| MFCD00006919 | |
| C.I. 26150, Fat Black HB, Solvent Black 3 | |
| (2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene |
Specifications
| 120.0°C to 124.0°C | |
| C29H24N6 | |
| C.I. 26150, Fat Black HB, Solvent Black 3 | |
| YCUVUDODLRLVIC-UHFFFAOYSA-N | |
| Authentic | |
| 456.553 | |
| 456.54 | |
| Complies | |
| Fine Crystalline Powder |
| 4197-25-5 | |
| MFCD00006919 | |
| Solubility in water: insoluble. | |
| CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C | |
| (2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene | |
| 61336 | |
| High Purity | |
| Black to Brown | |
| Sudan Black B, high purity biological stain |
RUO – Research Use Only