missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan Black B, high purity biological stain

Catalog No. AC190160250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC190160250 25 g Glass bottle
AC190161000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC190160250 Supplier Thermo Scientific Chemicals Supplier No. 190160250

CAS: 4197-25-5 | C29H24N6 | 456.553 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4197-25-5
Molecular Formula C29H24N6
Molecular Weight (g/mol) 456.553
MDL Number MFCD00006919
InChI Key YCUVUDODLRLVIC-UHFFFAOYSA-N
Synonym C.I. 26150, Fat Black HB, Solvent Black 3
PubChem CID 61336
IUPAC Name (2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene
SMILES CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C

Specifications

Melting Point 120.0°C to 124.0°C
Solubility Information Solubility in water: insoluble.
Infrared Spectrum Authentic
Formula Weight 456.54
Grade High Purity
Packaging Glass bottle
UV Spectrometry Complies
Color Black to Brown
Quantity 25 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Sudan Black B, high purity biological stain
Show More Show Less

RUO – Research Use Only