missing translation for 'onlineSavingsMsg'
Learn More

Sucrose For Enzymatic Assay Kit SCA20, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014711
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014711 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014711 Supplier Merck Emd Millipore Supplier No. S1174500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 57-50-1
Physical Form Neat
Linear Formula C12H22O11
MDL Number MFCD00006626
Melting Point 185°C to 187°C
Synonym alpha-D-Glc-(1->2)-beta-D-Fru; alpha-D-Glucopyranosyl beta-D-fructofuranoside; beta-D-Fructofuranosyl-alpha-D-glucopyranoside; D(+)-Saccharose; Sugar
Type Analytical Standard
Quantity 500 mg
Formula Weight 342.30
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less