Learn More
Chemical Identifiers
| CAS | 57-50-1 |
|---|---|
| Molecular Formula | C12H22O11 |
| Molecular Weight (g/mol) | 342.30 |
| MDL Number | MFCD00006626 |
| InChI Key | CZMRCDWAGMRECN-PWPRYFECNA-N |
| Synonym | sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand |
| PubChem CID | 5988 |
| ChEBI | CHEBI:17992 |
| IUPAC Name | (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol |
| SMILES | OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
S2500GM
|
Fisher Chemical
S2500GM |
500 g | Poly Bottle |
Each for $311.86
Case of 6 Each for $1,871.16
|
|
||||
|
S250
|
Fisher Chemical
S250 |
50 kg | Fiber Drum |
Each for $6,519.07
|
|
||||
Chemical Identifiers
| 57-50-1 | |
| 342.30 | |
| CZMRCDWAGMRECN-PWPRYFECNA-N | |
| 5988 | |
| (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol |
| C12H22O11 | |
| MFCD00006626 | |
| sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand | |
| CHEBI:17992 | |
| OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O |
Specifications
| Pass Test (BP/EP) | |
| 190°C | |
| 0.05% max. (NF) | |
| C12H22O11 | |
| +66.3 to +67.0° (+ or -) (BP/EP), 65.9° min. (NF) | |
| CZMRCDWAGMRECN-PWPRYFECNA-N | |
| OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O | |
| 342.30 | |
| CHEBI:17992 | |
| BP/EP/NF | |
| Sucrose |
| 57-50-1 | |
| 6.5 to 7.5 | |
| 0.1% max. (BP/EP) | |
| MFCD00006626 | |
| sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand | |
| Meets Requirements (BP/EP/NF) | |
| (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol | |
| 5988 | |
| 342.3 | |
| Solid |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.
NFPA
Health:1
Flammability:1
Instability:0