missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sucrose diacetate hexaisobutyrate
CAS: 27216-37-1 | C40H62O19
$130.99 - $444.86
Chemical Identifiers
| CAS | 27216-37-1 |
|---|---|
| Molecular Formula | C40H62O19 |
| MDL Number | MFCD00041859 |
| Synonym | SAIB |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6619530
|
Thermo Scientific Chemicals
J6619530 |
250 g |
Each for $130.99
|
|
|||||
|
AAJ6619536
|
Thermo Scientific Chemicals
J6619536 |
500 g |
Each for $227.73
|
|
|||||
|
AAJ66195A1
|
Thermo Scientific Chemicals
J66195A1 |
1 kg |
Each for $444.86
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 27216-37-1 | |
| C40H62O19 | |
| MFCD00041859 | |
| SAIB |
Specifications
| 27216-37-1 | |
| Colorless to Yellow | |
| 288°C | |
| 1.457 | |
| 250 g | |
| +49° (c=1 in Chloroform) | |
| Liquid |
| -7°C | |
| 1.146 | |
| C40H62O19 | |
| MFCD00041859 | |
| SAIB | |
| 846.91 | |
| Sucrose diacetate hexaisobutyrate |
Safety and Handling
EINECSNumber : 204-771-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only