accessibility menu, dialog, popup

UserName

CAS RN 27216-37-1

H3CCH3OO(R)O(S)OH3COO(R)O(R)OCH3O(R)OOCH3CH3(S)OOCH3CH3(R)OOCH3H3C(S)OOH3CH3C(R)OOCH3H3C

CAS RN 27216-37-1

IUPAC Name: [(2R,3R,4S,5S)-5-[(acetyloxy)methyl]-5-{[(2R,3R,4S,5R,6R)-6-[(acetyloxy)methyl]-3,4,5-tris[(2-methylpropanoyl)oxy]oxan-2-yl]oxy}-3,4-bis[(2-methylpropanoyl)oxy]oxolan-2-yl]methyl 2-methylpropanoate
Synonyms: sucrose acetate isobutyrate
Molecular Weight (g/mol): 846.92
Molecular Formula: C40H62O19
InChi Key: UVGUPMLLGBCFEJ-SWTLDUCYSA-N
SMILES: CC(C)C(=O)OC[C@H]1O[C@@](COC(C)=O)(O[C@H]2O[C@H](COC(C)=O)[C@@H](OC(=O)C(C)C)[C@H](OC(=O)C(C)C)[C@H]2OC(=O)C(C)C)[C@@H](OC(=O)C(C)C)[C@@H]1OC(=O)C(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)