missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sucralose, NF, 98-102%, Spectrum™ Chemical
S1416, 56038-13-2, C12H19Cl3O8
$2957.51 - $18399.17
Chemical Identifiers
| CAS | 56038-13-2 |
|---|---|
| Molecular Formula | C12H19Cl3O8 |
| Molecular Weight (g/mol) | 397.63 |
| MDL Number | MFCD03648615 |
| InChI Key | BAQAVOSOZGMPRM-QBMZZYIRSA-N |
| IUPAC Name | (2R,3R,4R,5R,6R)-2-{[(2R,3S,4S,5S)-2,5-bis(chloromethyl)-3,4-dihydroxyoxolan-2-yl]oxy}-5-chloro-6-(hydroxymethyl)oxane-3,4-diol |
| SMILES | OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18607074
|
Spectrum Chemical Mfg Cor
S1416500GM |
500 g | Amber Glass Bottle |
Each for $2,957.51
|
|
||||
|
18607075
|
Spectrum Chemical Mfg Cor
S141625KGBL |
2.5 kg | Poly Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18607076
|
Spectrum Chemical Mfg Cor
S141610KGBL |
10 kg | Poly Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Sucralose, NF is used as an artificial sweetener in phamaceutical formulations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 56038-13-2 | |
| 397.63 | |
| BAQAVOSOZGMPRM-QBMZZYIRSA-N | |
| OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl |
| C12H19Cl3O8 | |
| MFCD03648615 | |
| (2R,3R,4R,5R,6R)-2-{[(2R,3S,4S,5S)-2,5-bis(chloromethyl)-3,4-dihydroxyoxolan-2-yl]oxy}-5-chloro-6-(hydroxymethyl)oxane-3,4-diol |
Specifications
| 56038-13-2 | |
| 0.007 | |
| C12H19Cl3O8 | |
| 500 g | |
| BAQAVOSOZGMPRM-QBMZZYIRSA-N | |
| (2R,3R,4R,5R,6R)-2-{[(2R,3S,4S,5S)-2,5-bis(chloromethyl)-3,4-dihydroxyoxolan-2-yl]oxy}-5-chloro-6-(hydroxymethyl)oxane-3,4-diol | |
| 98 to 102% |
| 1 | |
| Amber Glass Bottle | |
| MFCD03648615 | |
| +84.0° to +87.5° | |
| OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl | |
| 397.63 | |
| NF |