missing translation for 'onlineSavingsMsg'
Learn More

Sucralose, NF, 98-102%, Spectrum™ Chemical

Catalog No. 18607074
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607074 500 g Amber Glass Bottle
18607075 2.5 kg Amber Glass Bottle
18607076 10 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18607074 Supplier Spectrum Chemical Mfg Cor Supplier No. S1416500GM

S1416, 56038-13-2, C12H19Cl3O8

Spectrum™ Chemical Sucralose, NF is used as an artificial sweetener in phamaceutical formulations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56038-13-2
Molecular Formula C12H19Cl3O8
Molecular Weight (g/mol) 397.63
MDL Number MFCD03648615
InChI Key BAQAVOSOZGMPRM-QBMZZYIRSA-N
IUPAC Name (2R,3R,4R,5R,6R)-2-{[(2R,3S,4S,5S)-2,5-bis(chloromethyl)-3,4-dihydroxyoxolan-2-yl]oxy}-5-chloro-6-(hydroxymethyl)oxane-3,4-diol
SMILES OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl

Specifications

CAS Max % 1
Ignition Residue 0.007
Packaging Amber Glass Bottle
Quantity 500 g
Specific Rotation +84.0° to +87.5°
Percent Purity 98 to 102%
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Sucralose