missing translation for 'onlineSavingsMsg'
Learn More

Succinylcholine chloride, 96%, Thermo Scientific™

Chemical Identifiers

CAS 71-27-2
Molecular Formula C14H30Cl2N2O4
Molecular Weight (g/mol) 361.3
InChI Key YOEWQQVKRJEPAE-UHFFFAOYSA-L
Synonym succinylcholine chloride, succinyldicholine chloride, suxamethonium chloride, midarine, scoline, succinylcholine dichloride, anectine, lysthenon, succinylforte, lysthenone
PubChem CID 22475
ChEBI CHEBI:61219
IUPAC Name trimethyl-[2-[4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl]oxyethyl]azanium;dichloride
SMILES C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C.[Cl-].[Cl-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC460110050
SDS View Documents
Thermo Scientific Chemicals
460110050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC460110250
View Documents
Thermo Scientific Chemicals
460110250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC460111000
SDS View Documents
Thermo Scientific Chemicals
460111000
100g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

71-27-2
361.3
succinylcholine chloride, succinyldicholine chloride, suxamethonium chloride, midarine, scoline, succinylcholine dichloride, anectine, lysthenon, succinylforte, lysthenone
CHEBI:61219
C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C.[Cl-].[Cl-]
C14H30Cl2N2O4
YOEWQQVKRJEPAE-UHFFFAOYSA-L
22475
trimethyl-[2-[4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl]oxyethyl]azanium;dichloride

Specifications

71-27-2
95% min. (on dry substance) (Argentometry)
5g
Solubility in water: soluble.
C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C.[Cl-].[Cl-]
361.3
CHEBI:61219
96%
Authentic
C14H30Cl2N2O4
succinylcholine chloride, succinyldicholine chloride, suxamethonium chloride, midarine, scoline, succinylcholine dichloride, anectine, lysthenon, succinylforte, lysthenone
YOEWQQVKRJEPAE-UHFFFAOYSA-L
trimethyl-[2-[4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl]oxyethyl]azanium;dichloride
22475
361.3
Succinylcholine chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 200-747-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only