CAS RN 71-27-2
IUPAC Name:
trimethyl[2-({4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl}oxy)ethyl]azanium dichloride
Synonyms:
succinylcholine chloride bis(succinyldichlorocholine) succinylcholine dichloride (anhydrous) anhydrous succinylcholine chloride suxamethonium chloride (anhydrous) chlorsuccinylcholin suxamethonium dichloride anhydrous succinyldicholine dichloride succinyldicholine dichloride (anhydrous) anhydrous suxamethonium chloride
Molecular Weight (g/mol):
361.3
Molecular Formula:
C14H30Cl2N2O4
InChi Key:
YOEWQQVKRJEPAE-UHFFFAOYSA-L
SMILES:
[Cl-].[Cl-].C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C
1
–
1
of
1
results
1
Promotions Available
Succinylcholine chloride, 96%
Succinylcholine chloride, 96%
CAS: 71-27-2 Molecular Formula: C14H30Cl2N2O4 Molecular Weight (g/mol): 361.3 InChI Key: YOEWQQVKRJEPAE-UHFFFAOYSA-L Synonym: succinylcholine chloride,succinyldicholine chloride,suxamethonium chloride,midarine,scoline,succinylcholine dichloride,anectine,lysthenon,succinylforte,lysthenone PubChem CID: 22475 ChEBI: CHEBI:61219 IUPAC Name: trimethyl-[2-[4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl]oxyethyl]azanium;dichloride SMILES: C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C.[Cl-].[Cl-]