missing translation for 'onlineSavingsMsg'
Learn More

Succinylcholine chloride, 96%

Catalog No. AC460111000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460111000 100 g
AC460110050 5 g
AC460110250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC460111000 Supplier Thermo Scientific Chemicals Supplier No. 460111000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 71-27-2 | C14H30Cl2N2O4 | 361.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71-27-2
Molecular Formula C14H30Cl2N2O4
Molecular Weight (g/mol) 361.3
InChI Key YOEWQQVKRJEPAE-UHFFFAOYSA-L
Synonym succinylcholine chloride, succinyldicholine chloride, suxamethonium chloride, midarine, scoline, succinylcholine dichloride, anectine, lysthenon, succinylforte, lysthenone
PubChem CID 22475
ChEBI CHEBI:61219
IUPAC Name trimethyl-[2-[4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl]oxyethyl]azanium;dichloride
SMILES C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C.[Cl-].[Cl-]

Specifications

Melting Point 160.0°C to 162.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (on dry substance) (Argentometry)
Quantity 100 g
Solubility Information Solubility in water: soluble.
Formula Weight 361.3
Percent Purity 96%
Chemical Name or Material Succinylcholine chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 200-747-4

RUO – Research Use Only