missing translation for 'onlineSavingsMsg'
Learn More

Succinonitrile 99.0+%, TCI America™

Catalog No. S0109500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0109500G 500 g
S010925G 25 g
S0109100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S0109500G Supplier TCI America Supplier No. S0109500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-61-2
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
MDL Number MFCD00001949
InChI Key IAHFWCOBPZCAEA-UHFFFAOYSA-N
Synonym succinonitrile, deprelin, succinodinitrile, ethylene cyanide, dician, dinile, 1,2-dicyanoethane, ethylene dicyanide, disuxyl, succinil
PubChem CID 8062
IUPAC Name butanedinitrile
SMILES N#CCCC#N

Specifications

Color White
Boiling Point 159°C
Quantity 500 g
UN Number 3439
Formula Weight 80.09
Percent Purity ≥99.0% (GC)
Physical Form Crystals
Chemical Name or Material Succinonitrile
EINECSNumber (2)-2771&(2)-1526
RTECSNumber WN3850000
TSCA Yes