accessibility menu, dialog, popup

UserName

NN

CAS RN 110-61-2

IUPAC Name: butanedinitrile
Synonyms: succinonitrile
Molecular Weight (g/mol): 80.09
Molecular Formula: C4H4N2
InChi Key: IAHFWCOBPZCAEA-UHFFFAOYSA-N
SMILES: N#CCCC#N
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)