Learn More
Thermo Scientific™ Succinimidyl acetate
Chemical Identifiers
| CAS | 14464-29-0 |
|---|---|
| Molecular Formula | C6H7NO4 |
| Molecular Weight (g/mol) | 157.13 |
| InChI Key | SIFCHNIAAPMMKG-UHFFFAOYSA-N |
| IUPAC Name | 2,5-dioxopyrrolidin-1-yl acetate |
| SMILES | CC(=O)ON1C(=O)CCC1=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC464750050
|
Thermo Scientific™
464750050 |
5 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC464750250
|
Thermo Scientific™
464750250 |
25 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC464751000
|
Thermo Scientific™
464751000 |
100 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 14464-29-0 | |
| 157.13 | |
| 2,5-dioxopyrrolidin-1-yl acetate |
| C6H7NO4 | |
| SIFCHNIAAPMMKG-UHFFFAOYSA-N | |
| CC(=O)ON1C(=O)CCC1=O |
Specifications
| 14464-29-0 | |
| 5 g | |
| SIFCHNIAAPMMKG-UHFFFAOYSA-N | |
| 2,5-dioxopyrrolidin-1-yl acetate | |
| 157.13 |
| C6H7NO4 | |
| Solubility aq. soln.: .. Other solubilities: soluble in toluene | |
| CC(=O)ON1C(=O)CCC1=O | |
| 157.13 | |
| Succinimidyl acetate |
Safety and Handling
GHS H Statement Causes skin irritation.
Causes serious eye irritation.
GHS P Statement IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 238-454-9
RUO â Research Use Only