Learn More
Thermo Scientific™ Succinimidyl acetate
Supplier: Thermo Scientific™ 464750250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Succinimidyl acetate | |
| C6H7NO4 | |
| Solubility aq. soln.: .. Other solubilities: soluble in toluene | |
| 157.13 |
| 14464-29-0 | |
| 25 g | |
| 157.13 |
Chemical Identifiers
| 14464-29-0 | |
| C6H7NO4 | |
| 157.13 |
Safety and Handling
GHS P Statement IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 238-454-9
RUO â Research Use Only