missing translation for 'onlineSavingsMsg'
Learn More

Succinimidyl 4-[3,5-Dimethyl-4-(4-nitrobenzyloxy)phenyl]-4-oxobutyrate 96.0+%, TCI America™

Catalog No. S0808100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0808100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0808100MG Supplier TCI America Supplier No. S0808100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 948995-62-8
Molecular Formula C23H22N2O8
Molecular Weight (g/mol) 454.435
InChI Key CUTCHWUKZOOSRX-UHFFFAOYSA-N
Synonym 4-[3,5-Dimethyl-4-(4-nitrobenzyloxy)phenyl]-4-oxobutyric Acid Succinimidyl Ester
PubChem CID 17751994
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-[3,5-dimethyl-4-[(4-nitrophenyl)methoxy]phenyl]-4-oxobutanoate
SMILES CC1=CC(=CC(=C1OCC2=CC=C(C=C2)[N+](=O)[O-])C)C(=O)CCC(=O)ON3C(=O)CCC3=O

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 454.44
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Succinimidyl 4-[3,5-Dimethyl-4-(4-nitrobenzyloxy)phenyl]-4-oxobutyrate
TSCA No
Recommended Storage Refrigerator