missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Succinimide 98.0+%, TCI America™
$41.02 - $132.35
Chemical Identifiers
| CAS | 123-56-8 |
|---|---|
| Molecular Formula | C4H5NO2 |
| Molecular Weight (g/mol) | 99.089 |
| MDL Number | MFCD00005495 |
| InChI Key | KZNICNPSHKQLFF-UHFFFAOYSA-N |
| Synonym | succinimide, 2,5-pyrrolidinedione, butanimide, succinic acid imide, 2,5-dioxopyrrolidine, 2,5-diketopyrrolidine, succinic imide, succinimide-sauba, 3,4-dihydropyrrole-2,5-dione, dihydro-3-pyrroline-2,5-dione |
| PubChem CID | 11439 |
| ChEBI | CHEBI:9307 |
| IUPAC Name | pyrrolidine-2,5-dione |
| SMILES | C1CC(=O)NC1=O |
Chemical Identifiers
| 123-56-8 | |
| 99.089 | |
| KZNICNPSHKQLFF-UHFFFAOYSA-N | |
| 11439 | |
| pyrrolidine-2,5-dione |
| C4H5NO2 | |
| MFCD00005495 | |
| succinimide, 2,5-pyrrolidinedione, butanimide, succinic acid imide, 2,5-dioxopyrrolidine, 2,5-diketopyrrolidine, succinic imide, succinimide-sauba, 3,4-dihydropyrrole-2,5-dione, dihydro-3-pyrroline-2,5-dione | |
| CHEBI:9307 | |
| C1CC(=O)NC1=O |
Specifications
| 123-56-8 | |
| White | |
| C4H5NO2 | |
| 25 g | |
| KZNICNPSHKQLFF-UHFFFAOYSA-N | |
| pyrrolidine-2,5-dione | |
| 11439 | |
| 99.09 | |
| Crystalline Powder |
| 125°C | |
| 130°C | |
| MFCD00005495 | |
| succinimide, 2,5-pyrrolidinedione, butanimide, succinic acid imide, 2,5-dioxopyrrolidine, 2,5-diketopyrrolidine, succinic imide, succinimide-sauba, 3,4-dihydropyrrole-2,5-dione, dihydro-3-pyrroline-2,5-dione | |
| C1CC(=O)NC1=O | |
| 99.089 | |
| CHEBI:9307 | |
| ≥98.0% (T) | |
| Succinimide |
Safety and Handling
EINECSNumber : (5)-0120
RTECSNumber : WN2200000
TSCA : Yes