missing translation for 'onlineSavingsMsg'
Learn More

Succinimide, 98+%

CAS: 123-56-8 | C4H5NO2 | 99.089 g/mol

$104.15 - $1198.64

Chemical Identifiers

CAS 123-56-8
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.089
MDL Number MFCD00005495
InChI Key KZNICNPSHKQLFF-UHFFFAOYSA-N
Synonym succinimide, 2,5-pyrrolidinedione, butanimide, succinic acid imide, 2,5-dioxopyrrolidine, 2,5-diketopyrrolidine, succinic imide, succinimide-sauba, 3,4-dihydropyrrole-2,5-dione, dihydro-3-pyrroline-2,5-dione
PubChem CID 11439
ChEBI CHEBI:9307
IUPAC Name pyrrolidine-2,5-dione
SMILES C1CC(=O)NC1=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1350330
View Documents
Thermo Scientific Chemicals
A1350330
250 g
Each for $104.15
Only null left
 
AAA135030B
View Documents
Thermo Scientific Chemicals
A135030B
1000 g
Each for $293.04
Only null left
 
AAA135030I
View Documents
Thermo Scientific Chemicals
A135030I
5000 g
Each for $1,198.64
Only null left
 
Description

Description

Succinimide is used in organic syntheses and in industrial silver plating processes. It is also used as anticonvulsant drugs such as ethosuximide, phensuximide and methsuximide. Further, it is used to form covalent bonds between proteins, peptides and plastics. In addition to this, it is used as an important raw material for the synthesis of heterocyclic compounds through N-acyliminium cyclizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Succinimide is used in organic syntheses and in industrial silver plating processes. It is also used as anticonvulsant drugs such as ethosuximide, phensuximide and methsuximide. Further, it is used to form covalent bonds between proteins, peptides and plastics. In addition to this, it is used as an important raw material for the synthesis of heterocyclic compounds through N-acyliminium cyclizations.

Solubility
Soluble in water and ethanol. Insoluble in ether and chloroform.

Notes
Incompatible with strong oxidizing agents, acids, bases and strong reducing agents.
Specifications

Chemical Identifiers

123-56-8
99.089
KZNICNPSHKQLFF-UHFFFAOYSA-N
11439
pyrrolidine-2,5-dione
C4H5NO2
MFCD00005495
succinimide, 2,5-pyrrolidinedione, butanimide, succinic acid imide, 2,5-dioxopyrrolidine, 2,5-diketopyrrolidine, succinic imide, succinimide-sauba, 3,4-dihydropyrrole-2,5-dione, dihydro-3-pyrroline-2,5-dione
CHEBI:9307
C1CC(=O)NC1=O

Specifications

123-56-8
1.41
287°C to 288°C
Odorless
MFCD00005495
108440
succinimide, 2,5-pyrrolidinedione, butanimide, succinic acid imide, 2,5-dioxopyrrolidine, 2,5-diketopyrrolidine, succinic imide, succinimide-sauba, 3,4-dihydropyrrole-2,5-dione, dihydro-3-pyrroline-2,5-dione
KZNICNPSHKQLFF-UHFFFAOYSA-N
pyrrolidine-2,5-dione
11439
99.09
Succinimide
121°C to 126°C
4 to 6
201°C (393°F)
C4H5NO2
250 g
14,8871
Soluble in water and ethanol. Insoluble in ether and chloroform.
C1CC(=O)NC1=O
99.089
CHEBI:9307
≥98%
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 204-635-6

RTECSNumber : WN2200000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only