Learn More
Succinimide, 98%
CAS: 123-56-8 | C4H5NO2 | 99.08 g/mol
$59.43 - $73.51
Chemical Identifiers
| CAS | 123-56-8 |
|---|---|
| Molecular Formula | C4H5NO2 |
| Molecular Weight (g/mol) | 99.08 |
| MDL Number | MFCD00005495 |
| InChI Key | KZNICNPSHKQLFF-UHFFFAOYSA-N |
| Synonym | succinimide, 2,5-pyrrolidinedione, butanimide, succinic acid imide, 2,5-dioxopyrrolidine, 2,5-diketopyrrolidine, succinic imide, succinimide-sauba, 3,4-dihydropyrrole-2,5-dione, dihydro-3-pyrroline-2,5-dione |
| PubChem CID | 11439 |
| ChEBI | CHEBI:9307 |
| IUPAC Name | pyrrolidine-2,5-dione |
| SMILES | C1CC(=O)NC1=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC348650050
|
Thermo Scientific Chemicals
348650050 |
5 g | Glass bottle |
Each for $73.51
|
|
||||
|
AC348651000
|
Thermo Scientific Chemicals
348651000 |
100 g | Glass bottle |
Each for $59.43
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 123-56-8 | |
| 99.08 | |
| KZNICNPSHKQLFF-UHFFFAOYSA-N | |
| 11439 | |
| pyrrolidine-2,5-dione |
| C4H5NO2 | |
| MFCD00005495 | |
| succinimide, 2,5-pyrrolidinedione, butanimide, succinic acid imide, 2,5-dioxopyrrolidine, 2,5-diketopyrrolidine, succinic imide, succinimide-sauba, 3,4-dihydropyrrole-2,5-dione, dihydro-3-pyrroline-2,5-dione | |
| CHEBI:9307 | |
| C1CC(=O)NC1=O |
Specifications
| 123-56-8 | |
| Brown to White | |
| 201°C | |
| 98% | |
| C4H5NO2 | |
| 5 g | |
| 15,9001 | |
| Solubility in water: 240g/L (20°C). Other solubilities: 1g/24 mL alcohol - 1g/5 mL alcohol (60°C), insoluble in ether and chloroform | |
| C1CC(=O)NC1=O | |
| 99.08 | |
| CHEBI:9307 | |
| 98% | |
| Succinimide, 98% |
| 121.0°C to 126.0°C | |
| 285.0°C to 290.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00005495 | |
| 21,369 | |
| succinimide, 2,5-pyrrolidinedione, butanimide, succinic acid imide, 2,5-dioxopyrrolidine, 2,5-diketopyrrolidine, succinic imide, succinimide-sauba, 3,4-dihydropyrrole-2,5-dione, dihydro-3-pyrroline-2,5-dione | |
| KZNICNPSHKQLFF-UHFFFAOYSA-N | |
| pyrrolidine-2,5-dione | |
| 11439 | |
| 99.08 | |
| Powder or Flakes |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap a
GHS Signal Word: Warning
EINECSNumber : 204-635-6
RTECSNumber : WN2200000
TSCA : TSCA
RUO – Research Use Only