missing translation for 'onlineSavingsMsg'
Learn More

Succinic acid, 99+%

CAS: 110-15-6 | C4H6O4 | 118.09 g/mol

Supplier:  Thermo Scientific Chemicals A120840C

 View more versions of this product

Catalog No. AAA120840C

Only null left
Explore available promotions

Description

Description

Succinic acid acts as a leaching agent in extracting lithium, cobalt from used Li-ion batteries and magnesium from magnesite ore. It is used as a starting material in the synthesis of new elastic polyesters, fumaric acid, succinic anhydride and alkyd resins. It plays an important role as an acidity regulator in the food and beverage industry, as an excipient in pharmaceutical products and acts as a cocrystallising agent in organic synthesis. Its derivative viz diethyl ester is used as a substrate in the Stobbe condensation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Succinic acid acts as a leaching agent in extracting lithium, cobalt from used Li-ion batteries and magnesium from magnesite ore. It is used as a starting material in the synthesis of new elastic polyesters, fumaric acid, succinic anhydride and alkyd resins. It plays an important role as an acidity regulator in the food and beverage industry, as an excipient in pharmaceutical products and acts as a cocrystallising agent in organic synthesis. Its derivative viz diethyl ester is used as a substrate in the Stobbe condensation.

Solubility
Soluble in ethanol, ethyl ether, acetone and methanol. Insoluble in toluene, benzene, carbon disulfide, carbon tetrachloride and petroleum ether.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
Specifications

Specifications

Succinic acid
110-15-6
C4H6O4
1754069
succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
KDYFGRWQOYBRFD-UHFFFAOYSA-N
butanedioic acid
1110
118.09
1.56
206°C (402°F)
186°C to 188°C
235°C
MFCD00002789
14,8869
Soluble in ethanol,ethyl ether,acetone and methanol. Insoluble in toluene,benzene,carbon disulfide,carbon tetrachloride and petroleum ether.
OC(=O)CCC(O)=O
118.09
CHEBI:15741
≥99%
10,000 g

Chemical Identifiers

110-15-6
118.09
KDYFGRWQOYBRFD-UHFFFAOYSA-N
1110
butanedioic acid
C4H6O4
MFCD00002789
succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
CHEBI:15741
OC(=O)CCC(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c

H318-H335

EINECSNumber : 203-740-4

RTECSNumber : WM4900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only