missing translation for 'onlineSavingsMsg'
Learn More

Succinic acid, 99+%

Catalog No. p-7081901
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10,000 g
2500 g
500 g
Catalog No. Quantity
AAA1208436 500 g
AAA120840E 2500 g
AAA120840C 10,000 g
3 options

Catalog No. AAA1208436

Supplier: Thermo Scientific Chemicals A1208436

Only null left

CAS: 110-15-6 | C4H6O4 | 118.09 g/mol

Succinic acid acts as a leaching agent in extracting lithium, cobalt from used Li-ion batteries and magnesium from magnesite ore. It is used as a starting material in the synthesis of new elastic polyesters, fumaric acid, succinic anhydride and alkyd resins. It plays an important role as an acidity regulator in the food and beverage industry, as an excipient in pharmaceutical products and acts as a cocrystallising agent in organic synthesis. Its derivative viz diethyl ester is used as a substrate in the Stobbe condensation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Succinic acid acts as a leaching agent in extracting lithium, cobalt from used Li-ion batteries and magnesium from magnesite ore. It is used as a starting material in the synthesis of new elastic polyesters, fumaric acid, succinic anhydride and alkyd resins. It plays an important role as an acidity regulator in the food and beverage industry, as an excipient in pharmaceutical products and acts as a cocrystallising agent in organic synthesis. Its derivative viz diethyl ester is used as a substrate in the Stobbe condensation.

Solubility
Soluble in ethanol, ethyl ether, acetone and methanol. Insoluble in toluene, benzene, carbon disulfide, carbon tetrachloride and petroleum ether.

Notes
Incompatible with bases, oxidizing agents and reducing agents.

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002789
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
Synonym succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
PubChem CID 1110
ChEBI CHEBI:15741
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(O)=O

Specifications

Melting Point 186°C to 188°C
Boiling Point 235°C
Beilstein 1754069
Merck Index 14,8869
Solubility Information Soluble in ethanol,ethyl ether,acetone and methanol. Insoluble in toluene,benzene,carbon disulfide,carbon tetrachloride and petroleum ether.
Formula Weight 118.09
Percent Purity ≥99%
Density 1.56
Quantity 500 g
Flash Point 206°C (402°F)
Chemical Name or Material Succinic acid
Show More Show Less
Hazard Category H318-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c
EINECSNumber 203-740-4
RTECSNumber WM4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only