missing translation for 'onlineSavingsMsg'
Learn More

Succinate, 0.2M buffer soln., pH 5.0, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals J63863AK

 View more versions of this product

Catalog No. AAJ63863AK

Only null left

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Succinate buffer soln.
Colorless
5.0
250 mL
Odorless
ZDQYSKICYIVCPN-UHFFFAOYSA-L
disodium;butanedioate
9020
150-90-3
0.2 M
Liquid
C4H4Na2O4
MFCD00002789
C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]
162.052
CHEBI:63675

Chemical Identifiers

150-90-3
162.052
ZDQYSKICYIVCPN-UHFFFAOYSA-L
CHEBI:63675
C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]
C4H4Na2O4
MFCD00002789
9020
disodium;butanedioate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H335

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions