missing translation for 'onlineSavingsMsg'
Learn More

Succinamide 98.0+%, TCI America™

Catalog No. S0096500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0096500G 500 g
S009625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0096500G Supplier TCI America Supplier No. S0096500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-14-5
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00008042
InChI Key SNCZNSNPXMPCGN-UHFFFAOYSA-N
Synonym succinamide, succindiamide, succinic amide, succinic acid diamide, succinic diamide, succinic acid diimide, acmc-1bnit, ethane-1,2-dicarboxamide, aronis25142
PubChem CID 8036
IUPAC Name butanediamide
SMILES C(CC(=O)N)C(=O)N

Specifications

Color White
Quantity 500 g
Formula Weight 116.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Succinamide
EINECSNumber (2)-0881
TSCA Yes