accessibility menu, dialog, popup

UserName

CAS RN 110-14-5

IUPAC Name: butanediamide
Synonyms: succinamide
Molecular Weight (g/mol): 116.12
Molecular Formula: C4H8N2O2
InChi Key: SNCZNSNPXMPCGN-UHFFFAOYSA-N
SMILES: NC(=O)CCC(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results