missing translation for 'onlineSavingsMsg'
Learn More

Succinaldehyde Disodium Bisulfite 92.0+%, TCI America™

Catalog No. S0216500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0216500G 500 g
S021625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0216500G Supplier TCI America Supplier No. S0216500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5450-96-4
Molecular Formula C4H8O8S2
Molecular Weight (g/mol) 248.22
MDL Number MFCD00024982
InChI Key SBHPIPPMYYFSHJ-ZXZARUISSA-L
Synonym sodium succinaldehyde bisulfite, succinaldehyde sodium bisulfite, succinaldehyde disodium bisulfite, succinaldehyde, bis sodium bisulfite, succinaldehyde-sodium bisulfite compound 1:2, succinaldehyde, disodium salt, 1, 1,4-dihydroxy-, disodium salt, wln: wsqyq2yqswq &-na-2
PubChem CID 54599174
IUPAC Name (1R,4S)-1,4-dihydroxybutane-1,4-disulfonate
SMILES O[C@@H](CC[C@@H](O)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Color White
Quantity 500 g
Formula Weight 294.20
Percent Purity ≥92.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Succinaldehyde Disodium Bisulfite
EINECSNumber (2)-0508&(2)-1599
RTECSNumber EK3585000
TSCA Yes
Recommended Storage Refrigerator