CAS RN 5450-96-4
CAS RN 5450-96-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Succinaldehyde Disodium Bisulfite 92.0+%, TCI America™
CAS: 5450-96-4 Molecular Formula: C4H8O8S2 Molecular Weight (g/mol): 248.22 MDL Number: MFCD00024982 InChI Key: SBHPIPPMYYFSHJ-ZXZARUISSA-L Synonym: sodium succinaldehyde bisulfite,succinaldehyde sodium bisulfite,succinaldehyde disodium bisulfite,succinaldehyde, bis sodium bisulfite,succinaldehyde-sodium bisulfite compound 1:2,succinaldehyde, disodium salt,1, 1,4-dihydroxy-, disodium salt,wln: wsqyq2yqswq &-na-2 PubChem CID: 54599174 IUPAC Name: (1R,4S)-1,4-dihydroxybutane-1,4-disulfonate SMILES: O[C@@H](CC[C@@H](O)S([O-])(=O)=O)S([O-])(=O)=O