missing translation for 'onlineSavingsMsg'
Learn More

Succinaldehyde Bis(dimethyl Acetal) 98.0+%, TCI America™

Catalog No. S03715G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03715G 5 g
S037125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S03715G Supplier TCI America Supplier No. S03715G
Only null left

Chemical Identifiers

CAS 6922-39-0
Molecular Formula C8H18O4
Molecular Weight (g/mol) 178.23
MDL Number MFCD00127971
InChI Key BJRGNROMZGSJDC-UHFFFAOYSA-N
Synonym Succinaldehyde Tetramethyl Acetal, 1,1,4,4-Tetramethoxybutane
PubChem CID 11263832
IUPAC Name 1,1,4,4-tetramethoxybutane
SMILES COC(CCC(OC)OC)OC

Specifications

Color Colorless
Boiling Point 89°C
Quantity 5 g
Formula Weight 178.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Succinaldehyde Bis(dimethyl Acetal)