accessibility menu, dialog, popup

UserName

CAS RN 6922-39-0

CH3OOH3COH3COCH3

CAS RN 6922-39-0

IUPAC Name: 1,1,4,4-tetramethoxybutane
Synonyms: 1,1,4,4-tetramethoxybutane
Molecular Weight (g/mol): 178.23
Molecular Formula: C8H18O4
InChi Key: BJRGNROMZGSJDC-UHFFFAOYSA-N
SMILES: COC(CCC(OC)OC)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results